Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sodium hydroxide solution 40% (w/w)

10 L
Catalog # APPC171220.0715
Corrosive
Supplier:   PanReac AppliChem
Sodium hydroxide solution 40% (w/w)
Catalog # APPC171220.0715
Supplier:  PanReac AppliChem
CAS Number:  1310-73-2
Sodium hydroxide solution 40% (w/w)

10 L

CAS Number:  1310-73-2
Price
378,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Absorbs CO₂ from air. Miscible with water.

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Density:1.43 g/ml
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

 

Specifications

Specification Test Results

  • Minimum Assay (Acidim.):
    40%
  • Nitrogen Compounds (as N):
    Max. 0.002%
  • Carbonate (as Na₂CO₃):
    Max. 1.0%

Specifications

  • Pk:
    10 L

Specifications