Sleeve samplers, U-D Master
Tip for U-D sleeve sampler, Stainless steel 316L, 0,1 ml
SAMP1500W-01
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
- Quick and easy accurate unit dose sampling
- Different volumes can be taken using the same sampler
- Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
- Volume:0,1 ml
- Description:Tip for U-D sleeve sampler
- Material:Stainless steel 316L
- Seq:06



