Order Entry
Export
Orders LinkContactUsLinkComponent
Bromophenol blue ACS

Bromophenol blue ACS

Catalog # GENORC-114
Supplier:   G-Biosciences
Bromophenol blue ACS
Catalog # GENORC-114
Supplier:  G-Biosciences
CAS Number:  115-39-9
Bromophenol blue ACS
CAS Number:  115-39-9
Price
1 490,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2