Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) 1 * 1 items
Catalog # BIND8012-1147
Supplier:
Binder
Expand image
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) 1 * 1 items
Catalog # BIND8012-1147
Supplier:
Binder