Stoppers for conical ground joints
14/23, Amber glass
Catalog # BRND144673
Supplier: Brand
Jump to:
Product Details & Documents
DURAN®. Hollow, with drip tip and hexagonal grip.
Certifications: DIN 12252.
Documents
BRND144635_TDS_EN
Datasheet
Specifications
- NS:14/23
- Type:Amber glass
- Seq:14