Orders LinkContactUsLinkComponent
Stoppers for conical ground joints
Stoppers for conical ground joints
10/19, Amber glass
Catalog # BRND144671
Supplier:   Brand
Stoppers for conical ground joints
Catalog # BRND144671
Supplier:  Brand
Stoppers for conical ground joints
Stoppers for conical ground joints
10/19, Amber glass
Price
7,10 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®. Hollow, with drip tip and hexagonal grip.
Certifications: DIN 12252.
Documents
Specifications
  • NS:
    10/19
  • Type:
    Amber glass
  • Seq:
    12