Order Entry
Export
Orders LinkContactUsLinkComponent
Scrapers, SteriWare®

Scrapers, SteriWare®

Sterile, 190 mm, 75×75 mm
Catalog # SAMP8785H-75S
Supplier:   Sampling Systems
Scrapers, SteriWare®
Catalog # SAMP8785H-75S
Supplier:  Sampling Systems
Scrapers, SteriWare®
Scrapers, SteriWare®

Sterile, 190 mm, 75×75 mm

Price
392,00 €
Log in for availability
1 * 100 items
Product Details & Documents

These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.

  • Moulded & packed in a cleanroom
  • Made from FDA and EU conforming HDPE
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10-6)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling

Full batch traceability

 : Individually bagged.

Specifications
  • Wiping area app.:
    75×75 mm
  • Description:
    Sterile
  • Length:
    190 mm
  • Seq:
    02