SwitchBox and 3-way ball valve, SymLine®
3-way ball valve, electrically operated, stainless steel, 3× GL 25 (m)
Catalog # SCTV160178
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Automatically divert waste liquids into the reserve canister.
- Switchbox compatible to all electronic S.C.A.T. Europe and SymLine® level sensors
- Also compatible with the Signalbox and Built-in signalbox
- Many more applications, not only for waste disposal of HPLC systems
Improved safety through increased collecting volume, the reserve canister protects against overflowing, even for a longer time period. For example, waste can be disposed of over the weekend without operator supervision and without having to empty or change the full container.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Description:3-way ball valve, electrically operated, stainless steel, 3× GL 25 (m)
- Seq:03



