Reagent troughs
Disposable reagent trough, 125 ml, Translucent
Catalog # TCAN10619196
Supplier: Tecan
Jump to:
Product Details & Documents
These disposable PP troughs are designed to optimise the performance and productivity of Tecan liquid handling platforms.
Specifications
- Capacity:125 ml
- Colour:Translucent
- Description:Disposable reagent trough
- Seq:04