Group lockout boxes for wall-mounting
Metal group lockout box, small, for 7 padlocks, Red, 203×57,15×178 mm
Catalog # BRDY105714
Supplier: Brady
Jump to:
Product Details & Documents
These lockout boxes are made of durable, powder-coated steel with key slot on top.
- Clear Lexan window slides out when locks are removed
- Comes with 7 or 15 key pegs that hold 5 keys each and ID labels to simplify key management
- Pre-drilled holes and hardware included for quick mounting
Legend language: English
Specifications
- Type:Metal group lockout box, small, for 7 padlocks
- Colour:Red
- Seq:01
- W×D×H:203×57,15×178 mm