Orders LinkContactUsLinkComponent
Labels for BradyJet J5000 lab printer
Labels for BradyJet J5000 lab printer
Continuous label printer tape, B-2595, J50C-6000-2595, Vinyl, Inkjet, White, BradyJet J5000, 152 mm×30 m
  BRDY148666
 :   Brady
Labels for BradyJet J5000 lab printer
  BRDY148666
 :  Brady
Labels for BradyJet J5000 lab printer
Labels for BradyJet J5000 lab printer
Continuous label printer tape, B-2595, J50C-6000-2595, Vinyl, Inkjet, White, BradyJet J5000, 152 mm×30 m
413,00 €
Log in for availability
1 * 1 Roll
This indoor marking film is an aqueous, inkjet printable, flexible, matte finish, pressure sensitive adhesive product that is designed to be used with the BradyJet™ J5000 colour label printer.
  • Specially-formulated topcoat enhances colours and label appearance
  • Topcoat protects print from smudging and harsh conditions
Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting.
Specifications
  • Printer model:
    BradyJet J5000
  • Printing method:
    Inkjet
  • Type:
    B-2595
  • Colour:
    White
  • Description:
    Continuous label printer tape
  • Dimensions:
    152 mm×30 m
  • Material:
    Vinyl
  • Seq:
    12
  • Model:
    J50C-6000-2595