Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
SOLVENT TRAP PTFE 1 * 1 items
SOLVENT TRAP PTFE 1 * 1 items
Print
SOLVENT TRAP PTFE 1 * 1 items
Catalog # HAAK222-1515
Supplier:
HAAKE
SOLVENT TRAP PTFE 1 * 1 items
Catalog # HAAK222-1515
Supplier:
HAAKE
Expand image
Return to Top
SOLVENT TRAP PTFE 1 * 1 items
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Return Policy
Product Details & Documents