ContactUsLinkComponent
This gene encodes a member of the B-cell receptor associated protein 31 superfamily. The encoded protein is a multi-pass transmembra ne protein of the endoplasmic reticulum that is involved in the anterograde transport of membrane proteins. 1 * 100 µG
  BWRLBS8676
This gene encodes a member of the B-cell receptor associated protein 31 superfamily. The encoded protein is a multi-pass transmembra ne protein of the endoplasmic reticulum that is involved in the anterograde transport of membrane proteins. 1 * 100 µG
  BWRLBS8676