Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CHIRALPAK® IG-3 150X2.1 MM 3ΜM ANALYTICAL COLUMN 1 * 1 items
Catalog # DAIC87594
Supplier:
CHIRAL TECHNOLOGIES
Expand image
CHIRALPAK® IG-3 150X2.1 MM 3ΜM ANALYTICAL COLUMN 1 * 1 items
Catalog # DAIC87594
Supplier:
CHIRAL TECHNOLOGIES