Order Entry
Export
Orders LinkContactUsLinkComponent
Vydac 208TP C8, HPLC columns

Vydac 208TP C8, HPLC columns

Guard kit for 2,1 mm ID columns, includes 1× guard cartridge, holder and coupler, Guard cartridge, C8, Stainless steel, 300 Å, 5 µm, Silica
Catalog # 208GK52/N
   Low Manufacturing Impact
Supplier:   Avantor
Vydac 208TP C8, HPLC columns
Catalog # 208GK52/N
Supplier:  Avantor
   Low Manufacturing Impact
Vydac 208TP C8, HPLC columns
Vydac 208TP C8, HPLC columns

Guard kit for 2,1 mm ID columns, includes 1× guard cartridge, holder and coupler, Guard cartridge, C8, Stainless steel, 300 Å, 5 µm, Silica

Price
494,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.

  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications

208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20K MW.

Vydac is a registered trademark of W.R.Grace and Co.

Specifications
  • Column Format:
    Guard cartridge
  • Particle Size:
    5 µm
  • Pore size:
    300 Å
  • Description:
    Guard kit for 2,1 mm ID columns, includes 1× guard cartridge, holder and coupler
  • Seq:
    47
  • Column hardware:
    Stainless steel
  • Phase:
    C8
  • Packing material:
    Silica
  • Cat. No.:
    208GK52/N
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Environmentally Preferable:
    Low Manufacturing Impact