ContactUsLinkComponent
Low-maintenance pH combination electrode, glass shaft, SILAMID® reference, 2 hole junction, DuraLid reference system, cylinder membr ane, A glass, screw plug head, length 120 mm, 12 mm Ø, 0...14 pH, 0...100 °C 1 * 1 items
Catalog # SCOI285113150
Supplier:  SI Analytics
Low-maintenance pH combination electrode, glass shaft, SILAMID® reference, 2 hole junction, DuraLid reference system, cylinder membr ane, A glass, screw plug head, length 120 mm, 12 mm Ø, 0...14 pH, 0...100 °C 1 * 1 items
Catalog # SCOI285113150
Supplier:  SI Analytics