Instrument trays
395×275×11 mm
Catalog # SAMPA674-395
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Made from 304 stainless steel.
- Good chemical resistance, very good temperature resistance
- Crevice free design
- High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.
Documents
SAMPA674-215_TDS_EN
Datasheet
Specifications
- Seq:03
- W×D×H:395×275×11 mm