Orders LinkContactUsLinkComponent
Adapters for capillary columns
Adapters for capillary columns
Siltek® treated adaptor for capillary column on split/splitless Injector
Catalog # REST20544
Supplier:   Restek
Adapters for capillary columns
Catalog # REST20544
Supplier:  Restek
Adapters for capillary columns
Adapters for capillary columns
Siltek® treated adaptor for capillary column on split/splitless Injector
Price
137,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These adapters are made of high quality stainless steel.
  • Ideal for Thermo Scientific TRACE and Focus SSL
  • Siltek®-treated version available for additional inertness
Specifications
  • Description:
    Siltek® treated adaptor for capillary column on split/splitless Injector