Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BUFFER SOLUTION PH12.45/ 0.05/ 25C 1 * 100 mL
BUFFER SOLUTION PH12.45/ 0.05/ 25C 1 * 100 mL
Print
BUFFER SOLUTION PH12.45/ 0.05/ 25C 1 * 100 mL
Catalog # REGEEP1245-100
Supplier:
REAGECON
BUFFER SOLUTION PH12.45/ 0.05/ 25C 1 * 100 mL
Catalog # REGEEP1245-100
Supplier:
REAGECON
Expand image
Return to Top
BUFFER SOLUTION PH12.45/ 0.05/ 25C 1 * 100 mL
Add to cart
Price
50,50 €
Log in
for availability
Unit of Measure
1 * 100 mL
-
+
Add to cart
Return Policy
Product Details & Documents