ContactUsLinkComponent
0.25MM I.D. DXI INTERNAL STANDARD TEE (PFA, GREEN MARKER). ATTACHES TO PORT #3 (DXI VERSION). SPECIALLY DESIGNED MIXING TEE IS PERFE CT FOR ONLINE ADDITION OF INTERNAL STANDARD OR ONLINE DILUTION. ACHIEVES EXCELLENT MIXING WITH MINIMAL DEAD VOLUME. 0 1 * 1 items
Catalog # PERCN0777662
Supplier:  PerkinElmer
0.25MM I.D. DXI INTERNAL STANDARD TEE (PFA, GREEN MARKER). ATTACHES TO PORT #3 (DXI VERSION). SPECIALLY DESIGNED MIXING TEE IS PERFE CT FOR ONLINE ADDITION OF INTERNAL STANDARD OR ONLINE DILUTION. ACHIEVES EXCELLENT MIXING WITH MINIMAL DEAD VOLUME. 0 1 * 1 items
Catalog # PERCN0777662
Supplier:  PerkinElmer