Order Entry
Export
Orders LinkContactUsLinkComponent
Ethanol absolute VLSI

Ethanol absolute VLSI

Plastic bottle, 5 L
Catalog # 85651.360
Flammable
Irritant
Supplier:   VWR Chemicals
Ethanol absolute VLSI
Catalog # 85651.360
Supplier:  VWR Chemicals
CAS Number:  64-17-5
Ethanol absolute VLSI
Ethanol absolute VLSI

Plastic bottle, 5 L

CAS Number:  64-17-5
Price
114,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78.3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.7895 g/cm³ (20 °C)
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications