Order Entry
Export
Orders LinkContactUsLinkComponent
Pierce™ Protein Concentrators

Pierce™ Protein Concentrators

Pierce™ Protein concentrator PES, 5 - 20 ml, 100 kD
Catalog # PIER88532
Pierce™ Protein Concentrators
Catalog # PIER88532
Pierce™ Protein Concentrators
Pierce™ Protein Concentrators

Pierce™ Protein concentrator PES, 5 - 20 ml, 100 kD

Price
175,00 €
Log in for availability
1 * 10 SET
Product Details & Documents

Protein Concentrators PES are disposable ultrafiltration centrifugal devices with a polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples such as proteins and nucleic acids.

  • Sample concentration of 10- to 30-fold can be achieved in 10 minutes or less
  • Unique design minimises membrane fouling and provides high speed concentrations, even with particle laden solutions
  • Compatible with most bench-top microcentrifuges with fixed-angle rotors that accommodate 2,2 ml tubes
  • Molecular weight cutoff
  • Can be used for processing sample volumes
  • Rapid protein concentration
  • Excellent protein recovery
  • Reliable, consistent results

The 0,5 ml concentrators contain a vertical PES membrane, which allows for processing of small volumes between 100 and 500 µl. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the retentate volume. Typical protein recovery for proteins that are ≥ two-fold greater than the MWCO is >90%, the minimum protein sample concentration should be 0,1 mg/ml.

The 2 to 6 ml concentrators have a built-in dead stop and contain a vertical low protein-binding, high flux PES membrane, which allows for processing of volumes between 2 and 6 ml. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the retentate volume.

Pierce™ Protein Concentrators PES contain a membrane rated for retaining molecules with a molecular weight at least twofold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications
  • MWCO:
    100 kD
  • Description:
    Pierce™ Protein concentrator PES, 5 - 20 ml
  • Seq:
    19
  • Cat. No.:
    PIER88532
  • Temperature:
    RT