Vydac 208TP C8, HPLC columns
Analytical, C8, Stainless steel, 300 Å, 5 µm, 2,10 mm, 250 mm, Silica
208TP52
: Avantor
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20K MW.
Vydac is a registered trademark of W.R.Grace and Co.
- Column Format:Analytical
- Ø int.:2,10 mm
- Particle Size:5 µm
- Pore size:300 Å
- Length:250 mm
- Seq:13
- Column hardware:Stainless steel
- Phase:C8
- Packing material:Silica
- Carbon load:5%
- Cat. No.:208TP52
- Environmental certification:ISO 14001:2015 (530840 UM15)
- Endcapped:Yes
- USP code:L7
- Environmentally Preferable:Low Manufacturing Impact



