Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY PLATE LID LOW PROFILE 96 CONDENSATION RINGS 1 * 100 items
Catalog # FTIT4TI-0282
Supplier:
4titude Ltd.
Expand image
ASSAY PLATE LID LOW PROFILE 96 CONDENSATION RINGS 1 * 100 items
Catalog # FTIT4TI-0282
Supplier:
4titude Ltd.