Orders LinkContactUsLinkComponent
Interchangeable blocks for thermo mixer
Interchangeable blocks for thermo mixer
Interchangeable tube block, SC-24C, 24×2 ml microtubes
Catalog # BSANBS-010143-EK
Supplier:   BIOSAN
Interchangeable blocks for thermo mixer
Catalog # BSANBS-010143-EK
Supplier:  BIOSAN
Interchangeable blocks for thermo mixer
Interchangeable tube block, SC-24C, 24×2 ml microtubes
Price
853,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Interchangeable tube block, SC-24C
  • Seq:
    02
  • For:
    24×2 ml microtubes