Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Bottle Assemblies, PC, Sterile, Single-Use

VWR® Bottle Assemblies, PC, Sterile, Single-Use

500 ml, 38-430, Port 1: PP Tube Plug
Port 2: 25 mm Hydrophobic PE Vent Filter, Port 1: 24"
Port 2: 3", C-Flex, Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4"
Catalog # 734-2854
Supplier:   VWR Collection
VWR® Bottle Assemblies, PC, Sterile, Single-Use
Catalog # 734-2854
Supplier:  VWR Collection
VWR® Bottle Assemblies, PC, Sterile, Single-Use
VWR® Bottle Assemblies, PC, Sterile, Single-Use

500 ml, 38-430, Port 1: PP Tube Plug
Port 2: 25 mm Hydrophobic PE Vent Filter, Port 1: 24"
Port 2: 3", C-Flex, Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4"

Price
1 210,00 €
Log in for availability
1 * 6 items
Product Details & Documents

Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.

  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6

Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

Specifications
  • Cap Size:
    38-430
  • Capacity:
    500 ml
  • Seq:
    03
  • Tubing connection:
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
  • Tubing length:
    Port 1: 24"
    Port 2: 3"
  • Tubing size:
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
  • Tubing material:
    C-Flex