ContactUsLinkComponent
Tegera Gloves 48, Chemical Protection, Nitrile 0.60mm, unflocked, diamond grip pattern, green, length 450mm, Cat III, AQL 0.65 1 * 1 PAIR
Catalog # EJEN48-11
Supplier:  EJENDALS
Tegera Gloves 48, Chemical Protection, Nitrile 0.60mm, unflocked, diamond grip pattern, green, length 450mm, Cat III, AQL 0.65 1 * 1 PAIR
Catalog # EJEN48-11
Supplier:  EJENDALS