Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thread adapter, GL PTFE , BOLA

With 2 GL threads, GL25 (from male thread) and GL45 (to male thread), 14,5 mm
  BOHLD537-16
 :   Bohlender
Thread adapter, GL PTFE , BOLA
  BOHLD537-16
 :  Bohlender
Thread adapter, GL PTFE , BOLA
Thread adapter, GL PTFE , BOLA

With 2 GL threads, GL25 (from male thread) and GL45 (to male thread), 14,5 mm

 
97,50 €
   
1 * 1 items
 

Straight transition thread adapter made of PTFE with two GL-threaded connections.

  • Chemical resistant
  • Vacuum suitable
  • Temperature resistant from –200 to +250 °C
 
  • Hole Ø:
    14,5 mm
  • Description:
    With 2 GL threads, GL25 (from male thread) and GL45 (to male thread)
  • Seq:
    04