pH/mV/°C/ISE meter, handheld, Eutech pH 450
pH/mV/ISE meter, handheld, Eutech pH 450 (Set 1)
662-2274
: EUTECH
This portable meter is ideal for use in any environment. It features a durable waterproof and dustproof design with IP67 rating.
- Large, backlit display with intuitive user-friendly icons
- Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
- Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
- Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
- Meets GLP standards: Built-in real-time clock stamps stored data and calibration data with time and date
- Can be used with two AA batteries or with optional universal power adapter (not included)
- Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter can be used with pH, ORP or ISE electrodes with BNC connector.
: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
- Description:pH/mV/ISE meter, handheld, Eutech pH 450 (Set 1)
- Seq:02
- ISE resolution:0,01 / 0,1 / 1
- Compliance:IP67
- ISE accuracy:±0,5 % full scale (monovalent); ±1 % full scale (divalent)
- Data logging capacity:500 data sets
- Power supply:Two AA batteries (included), AC adapter (optional)
- mV calibration:±200
- mV resolution:0,1 (±999,9); 1 (beyond)
- ISE calibration:From 2 - 4 or 2 - 5 points: 0,10 / 1,00 / 10,0 / 100,0 / 1000
0,20 / 2,00 / 20,0 / 200,0 / 2000
0,50 / 5,00 / 50,0 / 500,0 - W×D×H:200×83×57 mm
- Temperature range:−10,0...+110,0 °C
- ISE range:0,0 - 2000 ppm
- Temperature calibration:Offset in 0,1 increments, ±10 °C
- mV range:±2000
- pH range:−2,00...+16,00
- Temperature compensation:Automatic or manual from −10 to +110 °C
- pH calibration:Up to 5 points (USA & NIST & CUSTOM); up to 6 points (DIN)
- Temperature resolution:0,1 °C
- Temperature accuracy:±0,5 °C
- Weight:0,5 kg
- Cat. No.:662-2274
- pH accuracy:±0,01
- pH resolution:0,01
- Model:pH 450
- Outputs:USB, RS232
- mV accuracy:±0,2 +2 LSD or +0,5 % of reading; whichever is greater
- Display:Custom LCD with backlight



