Order Entry
Export
Orders LinkContactUsLinkComponent
pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/ISE meter, handheld, Eutech pH 450 (Set 3)
Catalog # 662-2312
Supplier:   EUTECH
pH/mV/°C/ISE meter, handheld, Eutech pH 450
Catalog # 662-2312
Supplier:  EUTECH
pH/mV/°C/ISE meter, handheld, Eutech pH 450
pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/ISE meter, handheld, Eutech pH 450 (Set 3)

Price
812,90 €
Log in for availability
1 * 1 items
Product Details & Documents

This portable meter is ideal for use in any environment. It features a durable waterproof and dustproof design with IP67 rating.

  • Large, backlit display with intuitive user-friendly icons
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
  • Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
  • Meets GLP standards: Built-in real-time clock stamps stored data and calibration data with time and date
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage

The meter can be used with pH, ORP or ISE electrodes with BNC connector.

Delivery information: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, USB cable, pH buffers, storage solution and rinse solution.

Documents

Specifications
  • Description:
    pH/mV/ISE meter, handheld, Eutech pH 450 (Set 3)
  • Seq:
    04
  • ISE resolution:
    0,01 / 0,1 / 1
  • Compliance:
    IP67
  • ISE accuracy:
    ±0,5 % full scale (monovalent); ±1 % full scale (divalent)
  • Data logging capacity:
    500 data sets
  • Power supply:
    Two AA batteries (included), AC adapter (optional)
  • mV calibration:
    ±200
  • mV resolution:
    0,1 (±999,9); 1 (beyond)
  • ISE calibration:
    From 2 - 4 or 2 - 5 points: 0,10 / 1,00 / 10,0 / 100,0 / 1000
    0,20 / 2,00 / 20,0 / 200,0 / 2000
    0,50 / 5,00 / 50,0 / 500,0
  • W×D×H:
    200×83×57 mm
  • Temperature range:
    −10,0...+110,0 °C
  • ISE range:
    0,0 - 2000 ppm
  • Temperature calibration:
    Offset in 0,1 increments, ±10 °C
  • mV range:
    ±2000
  • pH range:
    −2,00...+16,00
  • Temperature compensation:
    Automatic or manual from −10 to +110 °C
  • pH calibration:
    Up to 5 points (USA & NIST & CUSTOM); up to 6 points (DIN)
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,5 °C
  • Weight:
    0,5 kg
  • Cat. No.:
    662-2312
  • pH accuracy:
    ±0,01
  • pH resolution:
    0,01
  • Model:
    pH 450
  • Outputs:
    USB, RS232
  • mV accuracy:
    ±0,2 +2 LSD or +0,5 % of reading; whichever is greater
  • Display:
    Custom LCD with backlight