Orders LinkContactUsLinkComponent
Labels, hazard warning, electricity
Labels, hazard warning, electricity
Hazard labels, PIC 329-TRI 100-B7541, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Catalog # BRDY250288
Supplier:   Brady
Labels, hazard warning, electricity
Catalog # BRDY250288
Supplier:  Brady
Labels, hazard warning, electricity
Labels, hazard warning, electricity
Hazard labels, PIC 329-TRI 100-B7541, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Price
9,70 €
Log in for availability
1 * 1 items
Product Details & Documents
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
  • Self-adhesive
  • Clear identification of voltage, phases, grounding and insulation
Specifications
  • Colour:
    Black on yellow
  • Description:
    Hazard labels
  • Dimensions:
    100×87 mm
  • Material:
    Polyester
  • Seq:
    06
  • Model:
    PIC 329-TRI 100-B7541
  • Packed:
    3 labels/card