Novec™ 7100 engineered fluid
Poly bottle, 1 L
SHH0002-1L
Novec™ 7100 fluid consists of two inseparable isomers ((CF₃)₂CFCF₂OCH₃) and CF₃CF₂CF₂CF₂OCH₃) with essentially identical properties. Ideal for vapor degreasing applications as a neat (pure), azeotropic component and co-solvent parts cleaner.
- Non-flammable and non-conductive
- Compatible with plastics
- Low toxicity and surface tension
- Low global warming potential (GWP) and zero ozone depletion potential
- Histology Bath Fluid
Used as a heat transfer fluid, cleaning and rinsing agent, spray contact cleaner, cryofreezing solution, lubricant carrier, dielectric test media and replacement for CFC, HCFC, HFC and PFC.
Novec is a trademark of 3M Company
- :163702-08-7;163702-07-6
- :C₅H₃OF₉
- :MFCD28505595
- :250.06 g/mol
- Pack type:Poly bottle
- Pk:1 L