Order Entry
Export
Orders LinkContactUsLinkComponent
 

Novec™ 7100 engineered fluid

Poly bottle, 500 mL
Catalog # SHH0002-500ML
Novec™ 7100 engineered fluid
Catalog # SHH0002-500ML
CAS Number:  163702-08-7;163702-07-6
Novec™ 7100 engineered fluid
Novec™ 7100 engineered fluid

Poly bottle, 500 mL

CAS Number:  163702-08-7;163702-07-6
Price
194,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Novec™ 7100 fluid consists of two inseparable isomers ((CF₃)₂CFCF₂OCH₃) and CF₃CF₂CF₂CF₂OCH₃) with essentially identical properties. Ideal for vapor degreasing applications as a neat (pure), azeotropic component and co-solvent parts cleaner.

  • Non-flammable and non-conductive
  • Compatible with plastics
  • Low toxicity and surface tension
  • Low global warming potential (GWP) and zero ozone depletion potential
  • Histology Bath Fluid

Used as a heat transfer fluid, cleaning and rinsing agent, spray contact cleaner, cryofreezing solution, lubricant carrier, dielectric test media and replacement for CFC, HCFC, HFC and PFC.

Novec is a trademark of 3M Company

  • CAS Number:163702-08-7;163702-07-6
  • Formula:C₅H₃OF₉
  • MDL Number:MFCD28505595
  • Molecular Weight:250.06 g/mol
Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 mL

Specifications