Order Entry
Export
Orders LinkContactUsLinkComponent
E.Z.N.A.® Universal Pathogen DNA Kit

E.Z.N.A.® Universal Pathogen DNA Kit

E.Z.N.A.® Universal pathogen DNA kit, 5 Tests
Catalog # D4035-00
Supplier:   OMEGA BIO-TEK
E.Z.N.A.® Universal Pathogen DNA Kit
Catalog # D4035-00
Supplier:  OMEGA BIO-TEK
CAS Number:  
E.Z.N.A.® Universal Pathogen DNA Kit
E.Z.N.A.® Universal Pathogen DNA Kit

E.Z.N.A.® Universal pathogen DNA kit, 5 Tests

CAS Number:  
Price
28,20 €
Log in for availability
1 * 5 Tests
Product Details & Documents

The E.Z.N.A.® Universal Pathogen Kit allows for the rapid and reliable isolation of high quality host genomic DNA, gram positive and negative bacterial DNA, fungal spore DNA and viral DNA and viral RNA from tissue, urine, serum and fecal samples.

  • Isolates pathogen DNA & RNA from a variety of sample sources
  • Technology: Silica MicroElute spin column
  • Processing mode: Manual, centrifugation/vacuum
  • Starting amount: 25 to 30 mg tissue, 250 µl fecal sample/serum/urine/blood

This kit incorporates Omega Bio-tek’s Disruptor Tubes pre-filled with glass beads for faster and easier processing. The Disruptor Tubes allow for simultaneous homogenizaton and lysis of the samples in the kit’s lysis buffers and aid in effective lysis of difficult samples. No detergents are present in the initial lysis buffer, which eliminates foaming issues and provides optimal conditions for homogenization.

This unique buffer system does not require alcohol to bind nucleic acids, allowing for recovery of high quality DNA/RNA free of PCR inhibitors. Omega Bio-tek’s MicroElute LE Spin Columns are used, which allow for elution volumes as low as 15 µl.

Specifications

Specifications

  • Pk:
    5 Tests
  • Description:
    E.Z.N.A.® Universal pathogen DNA kit
  • Cat. No.:
    D4035-00
  • Prep size:
    Mini
  • Refine target molecule:
    Viral DNA|Bacterial DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)|Bead beating
  • Description:
    E.Z.N.A.® Universal pathogen DNA kit
  • Sample type:
    Stool|Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues

Specifications