Fractionating columns, Vigreux, Quickfit®
360 mm, 29/32 NS, 29/32 NS
BIBBFC3/14
PYREX®, borosilicate glass
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.
- Cone size:29/32 NS
- Socket size:29/32 NS
- Usable length:360 mm
- Seq:04