Orders LinkContactUsLinkComponent
Adapters with stopcock and socket
Adapters with stopcock and socket
24/29 NS, 3 mm
Catalog # BIBBMF14/3/SC
Supplier:   SciLabware
Adapters with stopcock and socket
Catalog # BIBBMF14/3/SC
Supplier:  SciLabware
Adapters with stopcock and socket
Adapters with stopcock and socket
24/29 NS, 3 mm
Price
521,00 €
Log in for availability
1 * 5 items
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .
Specifications
  • Hole Ø:
    3 mm
  • Socket size:
    24/29 NS