Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
250 ml, 29/32, 0,400 ± ml
Catalog # WITG3.670.025.29
Supplier:   witeg Labortechnik
Volumetric flasks, class A, white markings
Catalog # WITG3.670.025.29
Supplier:  witeg Labortechnik
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
250 ml, 29/32, 0,400 ± ml
Price
757,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
  • White markings
  • Conformity certified (DE-M marking)
  • Inclusive batch certificate
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    29/32
  • Capacity:
    250 ml
  • Seq:
    08
  • Tolerance:
    0,400 ± ml