Order Entry
Export
Orders LinkContactUsLinkComponent
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X
Catalog # ABCAAB193971-5
Supplier:   Abcam
Extraction Enhancer Buffer 50X, Abcam
Catalog # ABCAAB193971-5
Supplier:  Abcam
CAS Number:  
Extraction Enhancer Buffer 50X, Abcam

Extraction Enhancer Buffer 50X

CAS Number:  
Price
46,20 €
Log in for availability
1 * 5 mL
Product Details & Documents

Abcam's Extraction Enhancer Buffer 50X is formulated for the preparation of cell or tissues extracts for use in immunoassays.

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Extraction Enhancer Buffer 50X
  • Cat. No.:
    ABCAAB193971-5
  • Storage temperature:
    Store at +4°C.

Specifications