Order Entry
Export
Orders LinkContactUsLinkComponent
Human IgA ELISA Kit

Human IgA ELISA Kit

Human IgA ELISA kit, 96 tests
Catalog # ABCAAB196263-96
Supplier:   Abcam
Human IgA ELISA Kit
Catalog # ABCAAB196263-96
Supplier:  Abcam
CAS Number:  
Human IgA ELISA Kit
Human IgA ELISA Kit

Human IgA ELISA kit, 96 tests

CAS Number:  
Price
851,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human IgA ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantification of human Immunoglobulin A (IgA) with a sensitivity of 0,25 ng/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput. Broad sample compatibility - works with saliva urine, plasma, cell culture supernatants, milk, and serum.

Specifications

Specifications

  • Description:
    Human IgA ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Saliva, Urine, Plasma, Cell culture supernatant, Milk, Serum
  • Detection range:
    0,78 - 50 ng/ml
  • Storage temperature:
    4 ℃
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB196263-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,25 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    IgA
  • Regulatory status:
    RUO

Specifications