Cylinders, borosilicate glass, with hexagonal base
10 ml
Catalog # SCOT213910879
Supplier: DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Catalog # SCOT213910879
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
- Without graduation
- Hexagonal base
- Base with three knobs which increase its stability
: DIN EN ISO 4788.
Specifications
- Capacity:10 ml
- Seq:02