Orders LinkContactUsLinkComponent
Cylinders, borosilicate glass, with hexagonal base
Cylinders, borosilicate glass, with hexagonal base
5 ml
Catalog # SCOT213910776
Supplier:   DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Catalog # SCOT213910776
Supplier:  DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Cylinders, borosilicate glass, with hexagonal base
5 ml
Price
83,30 €
Log in for availability
1 * 10 items
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
  • Without graduation
  • Hexagonal base
  • Base with three knobs which increase its stability
Certifications: DIN EN ISO 4788.
Specifications
  • Capacity:
    5 ml