Order Entry
Export
Orders LinkContactUsLinkComponent
ASB-14 ≥97% zwitterionic detergent

ASB-14 ≥97% zwitterionic detergent

Catalog # ABCAAB142095-25
Supplier:   Abcam
ASB-14 ≥97% zwitterionic detergent
Catalog # ABCAAB142095-25
Supplier:  Abcam
CAS Number:  216667-08-2
ASB-14 ≥97% zwitterionic detergent
CAS Number:  216667-08-2
Price
820,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Zwitterionic detergent. Solubilizes proteins for 2D analysis, including membrane proteins.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Amidosulphobetaine-14
  • CAS Number:216667-08-2
  • Molecular Weight:434,7 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% ASB-14