0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
RECOMBINANT HUMAN MOCS2 PROTEIN 1 * 20 µG
ABCAAB134527-20
:
Abcam
RECOMBINANT HUMAN MOCS2 PROTEIN 1 * 20 µG
ABCAAB134527-20
:
Abcam