Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HUMAN SYCE3 PROTEIN 1 * 20 µG
Catalog # ABCAAB131666-20
Supplier:
Abcam
Expand image
RECOMBINANT HUMAN SYCE3 PROTEIN 1 * 20 µG
Catalog # ABCAAB131666-20
Supplier:
Abcam