0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
RECOMBINANT MOUSE AATK PROTEIN 1 * 20 µG
ABCAAB125617-20
:
Abcam
RECOMBINANT MOUSE AATK PROTEIN 1 * 20 µG
ABCAAB125617-20
:
Abcam