Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,9-Pyrazoloanthrone 99%

100 mg
Catalog # J67272.MC
Irritant
1,9-Pyrazoloanthrone 99%
Catalog # J67272.MC
CAS Number:  129-56-6
1,9-Pyrazoloanthrone 99%
1,9-Pyrazoloanthrone 99%

100 mg

CAS Number:  129-56-6
Price
214,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Purity: 99% CAS No.: 129-56-6 Molecular Formula: C14H8N2O Formula Weight: 220.23 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Yellow to brown Solubility: Soluble in DMSO at approx. 20mg/ml MDL No.: MFCD00022289

Synonyms:  JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
  • CAS Number:129-56-6
  • Formula:C₁₄H₈N₂O
  • MDL Number:MFCD00022289
  • Molecular Weight:220.231 g/mol
  • EINECS:204-955-6
Specifications

Specifications

  • Pk:
    100 mg

Specifications