Orders LinkContactUsLinkComponent
Combustion boats
Combustion boats
120×12×16 mm
Catalog # TSLA75147
Combustion boats
Catalog # TSLA75147
Combustion boats
120×12×16 mm
Price
98,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Seq:
    10
  • W×D×H:
    120×12×16 mm