Combustion boats
48×15×10 mm
Catalog # TSLA75143
Supplier: TECHNICAL QUARTZ SOLUTIONS
Jump to:
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifications
- Seq:06
- W×D×H:48×15×10 mm