Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Prepared Media for Microbiology
Dip slides
Dip slides
Print
Dip slides
Mikrocount® TPC, 20
Catalog # SCUE182812
Certificates
Supplier:
SCHUELKE MAYR
Dip slides
Catalog # SCUE182812
Supplier:
SCHUELKE MAYR
CAS Number:
Certificates
Expand image
Return to Top
Dip slides
Mikrocount® TPC, 20
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
51,24 €
Log in
for availability
Unit of Measure
1 * 20 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Specifications
Specifications
Pk:
20
Description:
Mikrocount® TPC
Specifications