PLOT columns, Rt®-Alumina BOND/MAPD (fused silica)
Capillary, 10,00 µm, 0,53 mm, 50 m
Catalog # REST19778
Supplier: Restek
Jump to:
Product Details & Documents
Rt-Alumina BOND/MAPD PLOT columns are made specifically for the analysis of petrochemicals and downstream products such as ethylene, propylene, butylenes, and butadiene.
- Optimised deactivation produces maximum response when analysing trace levels of acetylene, methyl acetylene, and propadiene
- Stable response factors make this column ideal for process-type applications where recalibration must be minimised
- High loadability reduces peak tailing and improves separations
- Extended temperature range up to 250 °C for fast elution of high molecular weight (HMW) hydrocarbons and accelerated column regeneration following exposure to water
Specifications
- Column Format:Capillary
- Film thickness:10,00 µm
- Ø int.:0,53 mm
- Length:50 m
- Seq:05



